Anti-BOD1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX51766
10162-070EA 988.07 CAD
10162-070
Anti-BOD1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to BOD1

Type: Primary
Antigen: BOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR