GelJacket™ Stackable CO₂ Incubators, 7404 Series, Caron Products
Supplier: Caron Products
7404-10-2 7404-10-3 7404-10-1
10158-770EA 41978.34 CAD
10158-770 10158-772 10158-852
GelJacket™ Stackable CO₂ Incubators, 7404 Series, Caron Products
Incubators CO₂ Incubators
Caron’s GelJacket is a high-volume, controlled humidity CO2 incubator that replaces conventional water jacketed units as the benchmark for critical culture protection.
GelJacket offers precise gVapor™ controlled humidification, as well as a rapid 90° decon cycle for fast chamber cleaning. These standard features save energy and user time, as opposed to contamination-prone water pan-based systems and high temperature or complicated competing decontamination processes.
Refrigerated models with extended temperature ranges are available for applications that generate heat (such as shakers).More Product Information