Anti-IgM µ Goat Polyclonal Antibody

Supplier: Immunoreagents
GTXHU-006-D
10152-008EA 235.97 CAD
10152-008
Anti-IgM µ Goat Polyclonal Antibody
Antibodies
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivityMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol IgM
Environmentally Preferable
Antigen name Immunoglobulin M mu-chain
Antibody type Secondary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Isotype IgM
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Epitope µ-chain
Immunogen Human IgM, µ chain
Format Purified IgG
Form Clear, colorless liquid, 0.2 µm filtered
Purification Affinity purified using solid phase Human IgM
Concentration > 4.5 mg/ml (E 1% at 280 nm = 13.0)
Cross adsorption No
Storage buffer 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Storage temperature 2-8 °C
Shipping temperature 2-8 °C
Tested applications Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR