Anti-IgG Dog Polyclonal Antibody

Supplier: Immunoreagents
DG-003-C5
10151-044EA 235.47 CAD
10151-044
Anti-IgG Dog Polyclonal Antibody
Antibodies
  • Origin: US

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Dog
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR