Anti-IgG Mouse Polyclonal Antibody

Supplier: Immunoreagents
MU-003-C
10151-034EA 464.38 CAD
10151-034
Anti-IgG Mouse Polyclonal Antibody
Antibodies
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR