Anti-IgG Goat Polyclonal Antibody (DyLight® 550)

Supplier: Immunoreagents
GTXRB-003-E550NHSX
10150-678EA 206.14 CAD
10150-678
Anti-IgG Goat Polyclonal Antibody (DyLight® 550)
Antibodies
The ImmunoReagents' DyLight® 550 conjugates provide vibrant orange-to-red fluorescence with better performance than other rhodamine derivatives, including Alexa Fluor 555, TRITC and Cy3 for fluorescent applications. DyLight® 550 conjugates have tight excitation/emission spectra that are ideal for multicolor applications. DyLight® 550 conjugated secondary antibodies are used in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 562nm / 576nm Emission Color = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3,More Product Information
Order Now

SPECIFICATIONS

Antigen symbol IgG
Antigen name Immunoglobulin G
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation DyLight® 550
Reactivity Rabbit
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Immunogen Rabbit IgG
Format Purified IgG
Form Lyophilized
Purification Affinity purified using solid phaseRabbit IgG
Concentration 1.0 mg/ml (E 1% at 280 nm = 13.0)
Cross adsorption Yes
Storage buffer 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
Storage temperature 2-8 °C
Shipping temperature 2-8 °C
Tested applications Fluoresence ELISA, Fluorescence-Activated Cell Sorting

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR