Anti-IgG F(c) Goat Polyclonal Antibody (DyLight® 650)

Supplier: Immunoreagents
GTXHU-004-F2650NHSX
10149-920EA 466.05 CAD
10149-920
Anti-IgG F(c) Goat Polyclonal Antibody (DyLight® 650)
Antibodies
  • Origin: US

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR