Anti-IgG Goat Polyclonal Antibody (DyLight® 633)

Supplier: Immunoreagents
GTXDK-003-D633NHSX
10149-668EA 289.09 CAD
10149-668
Anti-IgG Goat Polyclonal Antibody (DyLight® 633)
Antibodies
The ImmunoReagents' DyLight® 633 conjugates fluoresces red with physical properties comparable to other 633 dyes. DyLight® 633 is ideal for multi-color applications. DyLight® 633 is available as reactive labeling agents and as conjugates of secondary antibodies and biotin-binding proteins for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 638nm / 658nm Emission Color = Red (Similar Dyes: Alexa Fluor 633)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formatsMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol IgG
Antigen name Immunoglobulin G
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation DyLight® 633
Reactivity Donkey
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Immunogen Donkey IgG
Format Purified IgG
Form Lyophilized
Purification Affinity purified using solid phaseDonkey IgG
Concentration 1.0 mg/ml (E 1% at 280 nm = 13.0)
Cross adsorption No
Storage buffer 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
Storage temperature 2-8 °C
Shipping temperature 2-8 °C
Tested applications Fluoresence ELISA, Fluorescence-Activated Cell Sorting

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR