VWR® Ice Pans
Supplier: VWR International
10146-202 10146-204 10146-206 10146-208 10146-210 10146-212 10146-214 10146-216 10146-218 10146-220 10146-222 10146-224 10146-226 10146-228 10146-230 10146-232 10146-234 10146-236 10146-278 10146-280 10146-282 10146-294 10146-296 10146-298
10146-202EA 91.56 CAD
10146-202 10146-204 10146-206 10146-208 10146-210 10146-212 10146-214 10146-216 10146-218 10146-220 10146-222 10146-224 10146-226 10146-228 10146-230 10146-232 10146-234 10146-236 10146-278 10146-280 10146-282 10146-294 10146-296 10146-298
VWR® Ice Pans
Ice Containers
VWR® Ice Pans are manufactured from polyurethane foam to ensure good insulating properties.
  • Polyurethane foam construction
  • pouring lip for easy draining
  • Lids can be stacked beneath pans
Ice Pans are suitable for use with Dry-Ice with Isopropyl Alcohol, Water-Ice with salt slurry, or liquid nitrogen. The operating temperature range is -76 to 70°C (-104 to 158°F). All pans feature a pouring lip for easy draining. VWR® Ice Pans are available with or without lids. The lids, which are unbreakable in normal use, help to reduce evaporation levels. Lids canMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable