Sample Cylinder Valves (Stainless Steel & Sulfinert®-Treated), Restek
Supplier: Restek
21400 21401 21402-1 21402-69 21402-80 21402-11 21402-15 21402-17 21402-25 21402-29 21402-5 21402-68 26297 26298 26299 26300 26301-2 26301-6 26301-7 26301-9 26302 26303 26304 26305 26306 26307 26308 26309 26310 26311 26313-10 26313-4 26313-7 26314 26315 26316 26317 26301-1
10145-684EA 511.95 CAD
10145-684 10145-686 10145-688 10145-700 10145-722 10145-756 10145-758 10145-760 10145-768 10145-772 10145-790 10145-810 10145-864 10145-866 10145-868 10145-870 10145-872 10145-874 10145-878 10145-880 10145-884 10145-886 10145-888 10145-890 10145-892 10145-894 10145-896 10145-898 10145-900 10145-902 10145-904 10145-906 10145-912 10145-914 10145-916 10145-918 10145-920 10145-922 10145-924 10863-082
Sample Cylinder Valves (Stainless Steel & Sulfinert®-Treated), Restek
Cylinders Hydrometer Cylinders
Alta-Robbins' unique valve design incorporates a fully contained soft seat that provides durability and longer lifetime, and tight shut-off is easily achieved with very low torque, yet the valve is rugged enough to withstand overtightening.
  • Multiple valve configurations, including dip tube and rupture disks
  • Large, durable, Kel-F® seat ensures leak-free operation
  • Temperature range: -40°C to 120°C
  • 303 stainless steel
Multiple valve configurations are available for both high-pressure and ultra-high-pressure sample cylinders. An outage tube or dip tubeMore Product Information