Sample Cylinders, High Pressure (Stainless Steel & Sulfinert®-Treated), Restek
Supplier: Restek
21394 21394-PI 22921 24130 24130-PI 24131 24131-PI 24132 22921-PI 22922 22922-PI 22923 22923-PI 22924 22924-PI 22925 22925-PI 22926 22926-PI 24132-PI 24133 24133-PI 24134 24134-PI
10145-680EA 3896.18 CAD
10145-680 10145-682 10145-738 10145-746 10145-748 10145-750 10145-752 10145-754 10145-812 10145-814 10145-816 10145-818 10145-820 10145-822 10145-824 10145-826 10145-828 10145-830 10145-832 10145-844 10145-846 10145-848 10145-850 10145-852
Sample Cylinders, High Pressure (Stainless Steel & Sulfinert®-Treated), Restek
Cylinders Hydrometer Cylinders
Swagelok® sample cylinders are made of 304L and 316L stainless steel to resist corrosion and are DOT rated to 1,800 and 5,000 psig (TPED cylinders rated to 1,450 and 4,350 psig), which allows sampling at gas wellheads as well as on-site refineries.
  • 304L stainless steel; DOT rating to 1,800 psig (TPED cylinders to 1,450 psig)
  • Range of cylinder sizes, from 75 cc to 2,250 cc
  • All cylinders have 1/4" female NPT threads on both ends
  • TPED compliant cylinders available for EU community
Each cylinder is hydrostatically tested to at least 5/3 the working pressure.
Order Now
SPECIFICATIONS
Environmentally Preferable