Smart-Vue Remote Wireless Monitoring System, Thermo Scientific
Supplier: Thermo Fisher Scientific
SV103-501-LSB SV203-100-LSB SV208-200-LSB SV209-200-LSB SV200-200-LSB SV501-500-LSB SV501-502-LSB 89496-806 SV207-200-LSB SV300-500-LSB SV212-500-LSB SV100-500-LSB SV509-500-LSB SV204-100-LSB SV201-100-LSB SV202-100-LSB SV104-501-LSB SV204-200-LSB SV305-500-LSB SV401-200-LSB SV500-500-LSB SV101-500-LSB SV205-200-LSB SV401-100-LSB SV302-500-LSB SV208-100-LSB SV206-200-LSB 89496-886 89496-888 SV405-200-LSB SV201-200-LSB SV400-101-LSB SV209-100-LSB 89496-902 SV206-100-LSB SV207-100-LSB SV202-200-LSB SV501-501-LSB SV405-100-LSB SV303-500-LSB SV105-500-LSB SV204-300-LSB SV205-300-LSB 89496-934 SV603-500-LSB SV604-500-LSB SV605-500-LSB SV606-500-LSB SV607-500-LSB SV608-500-LSB SV609-500-LSB SV610-500-LSB SV611-500-LSB SV612-500-LSB
89496-778EA 2132.23 CAD
89496-778 89496-780 89496-782 89496-796 89496-798 89496-800 89496-802 89496-804 89496-806 89496-812 89496-814 89496-818 89496-822 89496-824 89496-826 89496-828 89496-830 89496-844 89496-846 89496-848 89496-850 89496-854 89496-856 89496-858 89496-862 89496-864 89496-866 89496-868 89496-870 89496-878 89496-880 89496-882 89496-886 89496-888 89496-890 89496-894 89496-898 89496-900 89496-902 89496-904 89496-906 89496-908 89496-910 89496-916 89496-920 89496-922 89496-924 89496-926 89496-930 89496-932 89496-934 10144-182 10144-184 10144-186 10144-188 10144-190 10144-192 10144-194 10144-196 10144-198 10144-200 10159-950 10159-952 10159-954 10159-956
Smart-Vue Remote Wireless Monitoring System, Thermo Scientific
Monitoring Systems CO2 and Temperature Monitoring Systems
This wireless monitoring solution for laboratory equipment safeguards the integrity of precious samples by continuously monitoring critical parameters and securely logging data to provide unprecedented peace-of-mind. Featuring audit trail traceability, this solution assists with conformance to 21 CFR Part 11, and other strenuous regulatory requirements for labs operating in regulated industries. System features software, modules, repeaters, receivers, and a 915 Mhz frequency (North America).
Continuous, 24/7/365 real-time monitoring of critical parameters, with intelligent data loggingMore Product Information