Silanized Glass Vials, Thermo Scientific
Supplier: Thermo Scientific
C39-20A/CT-S C39-20C/CT-S EP608154-S SCA-SV2-CT
10142-544CS 709.75 CAD
10142-544 10142-546 10142-548 10142-552
Silanized Glass Vials, Thermo Scientific
Vials
Save time and minimize waste costs when performing quantitative analysis or storing materials.
  • Trace organic analysis
  • Processing of materials prone to adhering to glass
  • Extraction glassware
  • Non-aqueous material storage
Surface deactivation treatments such as silanization may play an important role in preserving the integrity of certain materials or extracts stored in glass containers.
Pre-processed with a silanized surface deactivation treatment, these products are offered in convenient, ready-to-use clean packs.
It also helps to avoidMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable