Silicon carbide ≥98%, powder 18 µm

Supplier: Electron Microscopy Sciences
Danger

50260-18
101411-790EA 76.25 CAD
101411-790
Silicon carbide ≥98%, powder 18 µm
Silicon carbide

Hexagonal-alpha and beta phase silicone carbide for specimen preparation.


  • Dark grey 98% SiC
  • Moh's scale hardness is 9.0
  • Knoop Scale 2500


Available in 8, 10, 14, 18, 23, 35, 42, and 50 micron powder.

Formula: SiC
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR