Basic fuchsin stock solution 0.25% in water for Gram staining

Supplier: Electron Microscopy Sciences
Warning

26105-03
101411-032EA 72.85 CAD
101411-032
Basic fuchsin stock solution 0.25% in water for Gram staining
Basic Fuchsin

Brown and Brenn Method reagent for Gram Positive and Gram Negative Bacteria


Gram Negative Bacteria for bacteria stain: Gram + / Gram – bacteria and filaments of nocardia, etc


Stain results - Gram + Bacteria, Nocardia and Actinomyces Filaments: Blue, Gram – Bacteria, Nuclei: Red, Additional tissue elements: Yellow

Formula: C₂₀H₂₀ClN₃
MDL Number: MFCD00012569
CAS Number: 632-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR