Outdoor Labels for Moist, Frosty or Frozen Surfaces, Diversified Biotech
Supplier: Diversified Biotech
GRRL-1000
10141-760RL 117.47 CAD
10141-760
Outdoor Labels for Moist, Frosty or Frozen Surfaces, Diversified Biotech
Labels
Grips to wet, frost-covered and frozen surfaces; temperature range –80 to100 °C.
  • Thin material is compatible with centrifuge rotors
  • Resistant to chemicals and solvents
  • Adheres to plastics, glass, metals and even paperboard boxes
Choose your format: laser sheets for printing many labels at once, direct thermal labels for printing a few labels at a time or labels on a roll for handwriting labels.