Cryo Labels, Laser Printable, Diversified Biotech
Supplier: Diversified Biotech
CRY-2000-B CRY-2000-G CRY-2000-O CRY-2000-R CRY-2000-Y
10141-750RL 115.06 CAD
10141-750 10141-752 10141-754 10141-756 10141-758
Cryo Labels, Laser Printable, Diversified Biotech
Labels
Cryo-Tags® withstand both long term cryogenic storage conditions and conventional freezer storage.
  • Withstand flash/snap freezing in liquid nitrogen
  • General purpose
  • Temperature range between –196 to 79 °C
These labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks. It can withstand boiling water baths at 100 °C and dry heat up to 150 °C.
Order Now
SPECIFICATIONS
Environmentally Preferable