BRAND PCR Plates, BrandTech®
Supplier: Brandtech
781400 781354 781357 781412 781416 781345 781350 781411 781415 781366 781368 781371 781373 781375 781377 781367 781369 781372 781374 781376 781378
80087-150PK 908.18 CAD
80087-150 89135-444 89135-446 89135-456 89135-458 80087-142 89135-438 89400-702 89400-712 89400-714 10141-426 10141-428 10141-430 10141-432 10141-434 10141-436 10141-438 10141-440 10141-442 10141-444 10141-446 10141-448
BRAND PCR Plates, BrandTech®
Microplates PCR Plates
Designed for applications requiring a high level of accuracy and reproducibility, these 24-well, 48-well, 96-well, and 384-well PCR plates are constructed from high-quality virgin plastics without the addition of lubricants or chemical additives that can inhibit reactions and contaminate samples
  • Ultra-thin walls for optimal heat transfer
  • Cleanroom quality
  • Models to fit popular thermal cyclers
Ultra-thin walled design facilitates constant, rapid, and precise heat transfer. Smooth vessel interior minimizes the binding of enzymes and nucleic acid to theMore Product Information