Mouse IgG1 Isotype Control [clone: MOPC-21] (redFluor® 710)

Supplier: Tonbo Biosciences
80-4714-U100
10141-256EA 203.82 CAD
10141-256
Mouse IgG1 Isotype Control [clone: MOPC-21] (redFluor® 710)
Antibodies
The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Type: Control
Antigen: MOPC21
Clonality:
Clone: MOPC-21
Conjugation: redFluor® 710
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR