Anti-KLRB1C Mouse Antibody (redFluor® 710) [clone: PK136]
Supplier: Tonbo Biosciences
80-5941-U025 80-5941-U100
10141-126EA 102.7 CAD
10141-126 10141-128
Anti-KLRB1C Mouse Antibody (redFluor® 710) [clone: PK136]
Antibodies
The PK136 antibody is specific for mouse NK1.1, a type II transmembrane lectin-like receptor and member of the killer cell lectin-like receptor (KLR) family. NK1.1 is prominently expressed on natural killer (NK) cells, and is correlated with NK cytotoxic effects toward virus-infected cells and tumor cells. NK1.1 is expressed on subsets of NKT cells in certain mouse strains (C57BL/6, FVB/N, and NZB), yet absent from others (AKR, BALB/c, CBA/J, C3H, DBA/1, DBA/2, NOD, SJL, and 129). Putative subsets of NK cells and their expression of NK1.1 antigen are of continuing interest, including NK1.1+/More Product Information