Reverse Logistics Highway Transport Only Under 49 CFR 173.157 Marking Labels, Labelmaster
Supplier: Label Master
L500
101377-476EA 80.63 CAD
101377-476
Reverse Logistics Highway Transport Only Under 49 CFR 173.157 Marking Labels, Labelmaster
Labels
Help ensure compliance when shipping hazardous materials in limited quantities from a retail location back to the manufacturer, supplier, or distribution facility with the Reverse Logistics Highway Transport Markings.
  • Use on retail reverse logistics highway shipments of limited quantities shipped through a private carrier
  • Developed to help comply with 49 CFR 173.157
  • Printed on paper with outstanding chemical and abrasion resistance