Dulbecco's Modified Eagle's Medium (DMEM), Quality Biological

Supplier: QUALITY BIOLOGICAL, INC.

112-013-101 112-013-101CS 112-014-101 112-014-101CS 112-037-101 112-125-101 112-286-101
10128-206EA 78.49 CAD
10128-206 10128-208 10128-210 10128-212 10128-642 10128-650 10128-660
Dulbecco's Modified Eagle's Medium (DMEM), Quality Biological
Cell Culture Media

DMEM (Dulbecco’s Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells. We offer a variety of DMEM modifications for a range of cell culture applications.


  • With: High Glucose
  • Without: HEPES
  • Endotoxin and Mycoplasma tested


DMEM is one of the most widely used modifications of Basal Medium Eagle (BME). DMEM was originally developed as a richer version of BME; with 4 times higher concentrations of amino acids, vitamins and some additional components than BME. DMEM is commonly used for the culture of normal and transformed cells.


Recommendation
To DMEM without L-glutamine, it is recommended to add L-glutamine prior to use at a final concentration of 4.0 mM.


DMEM with high glucose supports increased cell growth. DMEM without phenol red is beneficial to downstream purification processing of expressed proteins (i.e. mAb, VLP) that require the use of various chromatography processes. The addition of sodium bicarbonate creates a high buffer capacity in the medium and keeps the pH-value in the physiological area during cultivation. DMEM: Ham's F-12 without L-glutamine contains no proteins, lipids or growth factors; and uses a sodium bicarbonate system (1.2 g/L), therefore requiring a 5 to 10% CO2 environment to maintain physiological pH. L-glutamine addition, to DMEM without L-glutamine, at the time of use allows for limited degradation over time.


QBI offers a variety of DMEM modifications for a range of cell culture applications.


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Caution: Research Use Only

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR