Water for molecular biology nuclease-, protease-free, low endotoxin, deionised, sterile

Supplier: QUALITY BIOLOGICAL, INC.

Synonyms: Hydrogen oxide

351-029-101 351-029-101CS 351-029-131 351-029-131CS 351-029-151 351-029-161 351-029-491 351-029-721 351-029-721EA
10128-452EA 57.84 CAD
10128-452 10128-454 10128-456 10128-458 10128-460 10128-462 10128-464 10128-466 10128-468
Water for molecular biology nuclease-, protease-free, low endotoxin, deionised, sterile
Water

Molecular Biology Water is prepared by reverse osmosis, passed through fine carbon, deionized through two resin beds, and serially filtered through 0.1 µm positively charged membranes.


  • DNase, RNase and protease tested


Molecular Biology Water is ideal for the preparation of reagents, rinsing glassware and plasticware, and other molecular biology applications where RNase, DNase, and Protease-free water is required.

No toxic agents, including DEPC, are used in the preparation of Molecular Biology Water, eliminating possible interferences with enzymatic reactions. Molecular Biology Water is negative for RNase (sensitivity to femtomoles) and is negative for non-specific Endonuclease, Exonuclease and Protease activity.

Molecular Biology Water is useful in: reconstituting nucleic acids and/or proteins, RNA applications (i.e. Preparation of an RNA probe), DNA applications (i.e. Subcloning in pUC plasmids), and protein applications (i.e. Western Blotting). Molecular Biology Water can also be used as a diluent or solvent for any molecular biology grade biochemical.

UNSPSC: 41116134
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: ambient


Caution: Research Use Only

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR