YPD/YEPD Plates, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
342-179-231
10128-392CS 1357.31 CAD
10128-392
YPD/YEPD Plates, Quality Biological
Media for Microbiology Prepared Media for Microbiology
YPD/YPED Plates are used for the growth of Saccharomyces cerevisiae (yeast).
A solid medium for the propagation of yeast.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only