YPG Plates, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
340-190-301
10128-348CS 1233.71 CAD
10128-348
YPG Plates, Quality Biological
Media for Microbiology Prepared Media for Microbiology
YPG Agar is recommended for the growth of Saccharomyces cerevisiae for molecular biology purpose.
  • YPG is a complex medium containing a nonfermentable carbon source (glycerol) that does not support the growth of ρ– or pet mutants.
YPG Plates are useful for isolation of Beta Carotene-Hyper producing Mutants and biocatalyst preparation.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: NA
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only