Intelli-Sense™ Multi-Speed Fiberglass Blowers, Labconco
Supplier: Labconco
7181820 7181822 7181811 7181821 7181813 7181823
10127-930EA 9148.2 CAD
10127-930 10128-044 76330-112 76330-114 76330-116 76330-118
Intelli-Sense™ Multi-Speed Fiberglass Blowers, Labconco
Ventilators
Intelli-Sense™ Multi-Speed Fiberglass Blowers are ideal for fume hood exhaust systems in moderate to highly corrosive conditions
They feature a direct drive ECM motor, which consumes one third less energy and is easier and less expensive to maintain than belt drive motors, and a phenolic-coated impeller and housing. Unit comes with control box and 3 speed blower switch. Adjustable over broad range of airflows.
The Intelli-Sense Blower provides an economical alternative to traditional on/off blowers. Users may close the sash and turn the Intelli-Sense Blower switch to standby mode soMore Product Information