RollerTool™ Adapter and Irradiated Covers, Micronova
Supplier: Micronova
QDRT-01 RTC1112-IR RTC2012-IRBB RTC612-IR
10126-354EA 678.48 CAD
10126-354 10126-356 10126-360 10126-362 10126-364
RollerTool™ Adapter and Irradiated Covers, Micronova
Mops
A custom mop with dual, curved plates, designed specifically for 3" roller bottle incubators, to clean upper and lower rollers at the same time. Can also be used for small piping and other cylindrical cleanroom surfaces.
  • Clean and sanitize roller bottle incubators with ease
  • Effectively cleans upper and lower rollers at the same time
  • Covers available in a choice of fabrics for specialized cleaning:
  • NovaPoly™ - smooth knit polyester provides excellent disinfectant dispersal
  • PolySorb™ - textured polyester for residue removal and dispersing disinfectant
More Product Information