EXP® Nano Fittings for 6-40 Ports and ¹/₃₂" Stainless Steel Tubing
Supplier: Optimize
15-19-04243 15-19-04247 15-19-04248 15-19-04249 15-19-04244 15-19-04245 15-19-04246 15-19-04242
10122-694PK 1458.06 CAD
10122-694 10122-776 10122-778 10122-780 10122-782 10122-784 10122-786 10122-788
EXP® Nano Fittings for 6-40 Ports and ¹/₃₂" Stainless Steel Tubing
Fittings and Connectors
The EXP Fitting System is the premier adjustable nut and ferrule compression fitting for extreme high-pressure connections between ¹/₃₂" or ¹/₁₆" tubing and any 6-32, 6-40 or 10-32 port.
  • Hand-Tight - rated to 20000+ psi (1400+ bar), tested to 30000 psi
  • Auto-adjusting ZDV Connection
  • Intended for many repeat uses
  • No more cut-off and replacement of swaged ferrules
  • Cost efficient with replaceable components
  • Simple and versatile - can be used in UHPLC and HPLC applications
The patented Titanium Hybrid Ferrule integrates the resilience of titanium with theMore Product Information