OPTI-TRAP™ Cartridges
Supplier: Optimize
10-04816-TO 10-04816-TP 10-04816-TM 10-04818-TN 10-04818-TO 10-04815-TN 10-04818-TP 10-04818-TM 10-04816-TN 10-04817-TP 10-04813-TP 10-04815-TP 10-04813-TM 10-04813-TN 10-04817-TO 10-04815-TO 10-04817-TM 10-04817-TN 10-04815-TM 10-04814-TN 10-04814-TP 10-04814-TM
10122-634PK 1768.08 CAD
10122-634 10122-636 10122-638 10122-640 10122-642 10122-644 10122-646 10122-648 10122-650 10122-652 10122-654 10122-656 10122-658 10122-660 10122-662 10122-664 10122-666 10122-668 10122-670 10122-672 10122-674 10122-676
OPTI-TRAP™ Cartridges
Chromatography Columns
The OPTI-TRAP™ is a bi-directional trap cartridge system which is used individually or in a series for sample concentration and purification.
  • For use up to 1500 psi (100 bar)
  • Low back pressure, for syringe loading/eluting or on-line trapping applications when plumbed into sample injection valve
  • Titanium and CTFE for chemical and biocompatibility
  • Transparent design allows for visual inspection of bed
  • Bidirectional for easy flushing and regeneration of the bed
  • Three bed sizes: 50, 5.0 and 0.5 µl
  • Color coded band identification for packing materials