VertiKlean® MAX™ Mop Wipe System
Supplier: Contec
VKMX0225 VKMX0275 VKSE0200 VKSE0250 2652K LWQT0100 VKSE0200B
89495-244CS 838.15 CAD
89495-244 10119-378 10171-158 10803-094 10816-268 76006-334 76006-338 77786-122
VertiKlean® MAX™ Mop Wipe System
Mops
The VertiKlean® MAX™ Mop Wipe System offers a quick and easy solution to mopping floors, walls, and other flat surfaces in any controlled environment. This versatile mop works with a variety of Contec® mop wipe covers, enabling the user to customize the mop to the cleaning task.
  • Stainless steel frame is chemical resistant
  • Autoclavable; can be sterilized by most common methods including gamma irradiation, ethylene oxide, or autoclave (recommended cycle is 121°C for 30 minutes)
  • Easy to attach and remove mop covers
  • Variety of fabric options available
  • Removes
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable