Epredia™ Microscope Slides and Cover Glasses
Supplier: Epredia
9991090 9991095 102460 152222 152250 152440 152450 152455 152460 202450 202460 12440S 12450S 12460S 9991001 9991002 9991003 9991004 9991009 9991011 9991012 9991013 9991014 6776214CS 4448 4450 4451 4440 4441 4442 4443 4444 4445 4446 4447
10034-098CS 264.11 CAD
10034-098 10034-100 CA16002-252 CA16002-256 CA16002-258 CA16002-262 CA16002-264 CA16002-266 CA16002-268 CA16002-314 CA16002-316 CA16002-530 CA16002-532 CA16002-534 10118-566 10118-568 10118-570 10118-572 10118-574 10118-576 10118-578 10118-580 10118-582 10118-584 10118-942 10118-944 10118-946 10118-948 10118-950 10118-952 10118-954 10118-956 10118-958 10118-960 10118-962
Epredia™ Microscope Slides and Cover Glasses
Slides Microscope Slides
Epredia™ offers a wide assortment of high-quality microscope slides and cover glass sizes that are available to meet the needs of modern laboratories.
  • Epredia™ cover glass and coverslips are uniquely manufactured to eliminate sticking and reduce waste
  • Made from the highest quality glass
  • Free from blisters and flaws, and consistent in cut, thickness and dimensional accuracy
  • Signature Series™ cover glass are an excellent choice for use with automatic coverslippers
Superfrost™ Plus and Colorfrost Plus™ microscope slides are positively charged forMore Product Information