Protector® PVC Perchloric Acid and Acid Digestion Laboratory Hoods, Labconco®
Supplier: Labconco
140410000 140410001 140410002 140410020 140410021 140510000 140510001 140510002 140510020 140510021 140610000 140610001 140610002 140610020 140610021 140810000 140810001 140810002 140810020 140810021 141410000 141410001 141410002 141410020 141410021 141510000 141510001 141510002 141510020 141510021 141610000 141610001 141610002 141610020 141610021 141810000 141810001 141810002 141810020 141810021
10118-468EA 45448.7 CAD
10118-468 10118-480 10118-482 10118-484 10118-486 10118-488 10118-490 10118-492 10118-494 10118-496 10118-498 10118-500 10118-502 10118-504 10118-506 10118-508 10118-510 10118-512 10118-514 10118-516 10118-518 10118-520 10118-522 10118-524 10118-526 10118-528 10118-530 10118-532 10118-534 10118-536 10118-538 10118-540 10118-542 10118-544 10118-546 10118-548 10118-550 10118-552 10118-554 10118-556
Protector® PVC Perchloric Acid and Acid Digestion Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Recommended whenever perchloric acid is used in combination with other corrosive inorganic reagents.
Interior and integral bottom with drainage trough are constructed of heat-welded Type I unplasticized polyvinyl chloride (PVC).
Perchloric acid hoods feature tempered safety glass sash. Acid digestion hoods utilize LEXAN® plastic sash to resist hydrofluoric acid exposure. Hoods have a glacier white powder-coated steel exterior.
Hoods are factory prepared for up to eight service fixtures and four electrical duplexes. They are supplied with two pre-plumbed service fixtures with forgedMore Product Information