Anti-VARS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15776-1-AP-150UL
10096-926EA 709.42 CAD
10096-926
Anti-VARS2 Rabbit Polyclonal Antibody
Antibodies
VARS2(Valine--tRNA ligase, mitochondrial) is also named as KIAA1885, VARS2L, VARSL and belongs to the class-I aminoacyl-tRNA synthetase family. The deduced 993-amino acid protein has an N-terminal mitochondrial targeting signal with a predicted cleavage site after residue 29. VARS2 has characteristics of a class I mitochondrial aminoacyl-tRNA synthetase, including a classical Rossmann fold.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: VARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR