Anti-ERP19 Rabbit Polyclonal Antibody

Supplier: Proteintech
20181-1-AP-150UL
10096-524EA 709.42 CAD
10096-524
Anti-ERP19 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: ERP19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR