Anti-TRAF3 Rabbit Polyclonal Antibody
Supplier: Proteintech
18099-1-AP-150UL
10095-712EA 709.42 CAD
10095-712
Anti-TRAF3 Rabbit Polyclonal Antibody
Antibodies
TNF receptor-associated factor 3 (TRAF3) has many alternative names including CAP-1, CD40 receptor-associated factor 1(CRAF1), CD40-binding protein(CD40BP) and LMP1-associated protein 1 (LAP1). TRAF3 regulates pathways leading to the activation of NF-kappa-B and MAP kinases, and plays a central role in the regulation of innate immune response. It modulates signaling pathways leading to the production of cytokines and interferon. As an essential constituent of several E3 ubiquitin-protein ligase complexes, TRAF3 promote 'Lys-63'-linked ubiquitination of target proteins. Moreover, TRAF3More Product Information