Anti-STX7 Rabbit Polyclonal Antibody

Supplier: Proteintech
12322-1-AP-150UL
10095-538EA 709.42 CAD
10095-538
Anti-STX7 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:1000-1:10000; IHC: Human medulloblastoma Tissue, 1:20-1:200; IP: Mouse Heart Tissue, 1:200-1:2000

Type: Primary
Antigen: STX7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR