Anti-SRR Rabbit Polyclonal Antibody

Supplier: Proteintech
17955-1-AP-150UL
10095-190EA 709.42 CAD
10095-190
Anti-SRR Rabbit Polyclonal Antibody
Antibodies
SRR(serine racemase) is a 37 kDa protein and belongs to the serine/threonine dehydratase family. It plays a role in neuronal activity and a potential biological role in the regulation of N-methyl-D-aspartate (NMDA) receptor activity in these peripheral organs as well. SRR catalyzes the racemization between l- and d-serine.

Western Blot:rat Brain Tissue, 1:200-1:1000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: SRR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR