Anti-PISD Rabbit Polyclonal Antibody

Supplier: Proteintech
16401-1-AP-150UL
10092-360EA 709.42 CAD
10092-360
Anti-PISD Rabbit Polyclonal Antibody
Antibodies
PISD(Phosphatidylserine decarboxylase proenzyme) belongs to the phosphatidylserine decarboxylase family.It contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.It has 2 isoforms produced by alternative splicing.This antibody is specific to PISD. The sequence correspond to the beta isoform.

Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: PISD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR