Anti-PMM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11252-2-AP-150UL
10092-292EA 709.42 CAD
10092-292
Anti-PMM1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: PMM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR