Anti-PIK3C2A Rabbit Polyclonal Antibody

Supplier: Proteintech
22028-1-AP-150UL
10092-266EA 709.42 CAD
10092-266
Anti-PIK3C2A Rabbit Polyclonal Antibody
Antibodies
Western Blot:PC-3 Cells, 1:500-1:5000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: PIK3C2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol PIK3C2A
Environmentally Preferable
Antigen name Phosphatidylinositol-4-Phosphate 3-Kinase, Catalytic Subunit Type 2 Alpha
Antigen synonyms DKFZp686L193|C2-containing phosphatidylinositol kinase|class 2|MGC142218|PI3K-C2alpha|CPK|phosphoinositide-3-kinase|PI3-K-C2A|PI3-K-C2(ALPHA)|alpha polypeptide|PIK3C2A
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunogen Fusion Protein
Purification Antigen affinity Purification
Storage buffer PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Shipping temperature Store at -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR