Anti-PGT Rabbit Polyclonal Antibody

Supplier: Proteintech
14327-1-AP-150UL
10092-170EA 709.42 CAD
10092-170
Anti-PGT Rabbit Polyclonal Antibody
Antibodies
Prostaglandin transporter (PGT), also known as SLCO2A1, has been identified as a functional prostaglandin (PG) carrier that mediates either the release of newly-synthesized PGs by cells and/or the uptake of PGs prior to intracellular oxidation. Northern blot analysis revealed that PGT mRNA has broad tissue distribution, suggesting its role in the transport and/ or metabolic clearance of PG in diverse tissues (8787677). This antibody is expected to detect the endogenous PGT protein.

Western Blot:NIH/3T3 Cells, 1:500-1:5000

Type: Primary
Antigen: PGT
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR