Anti-NR5A1 Rabbit Polyclonal Antibody
Supplier: Proteintech
18658-1-AP-150UL
10091-284EA 709.42 CAD
10091-284
Anti-NR5A1 Rabbit Polyclonal Antibody
Antibodies
Steroidogenic factor-1 (SF-1,STF-1), also known as NR5A1, regulates multiple genes involved in the adrenal and gonadal development and in the biosynthesis of a variety of hormones, including adrenal and gonadal steroids, anti-Mullerian hormone (AMH), and gonadotropins. SF-1 belongs to the fushi tarazu factor-1 (FTZ-F1) subfamily of orphan nuclear receptors. Initially identified as a tissue-specific transcriptional regulator of cytochrome P450 steroid hydroxylases, research studies of both global and tissue-specific knockout mice have demonstrated that SF-1 is required for the development ofMore Product Information