Anti-OXTR Rabbit Polyclonal Antibody

Supplier: Proteintech
23045-1-AP-150UL
10090-918EA 709.42 CAD
10090-918
Anti-OXTR Rabbit Polyclonal Antibody
Antibodies
Oxytocin (OXT) is a neurohypophysial hormone that plays a role in lactation and parturition and in the central nervous system as a neurotransmitter involved in sex and maternal behavior . Oxytocin acts through its receptor, OXTR, which belongs to the G protein-coupled 7-transmembrane receptor family. OXTR is expressed in endometrium, myometrium, mammary gland . This antibody is raised against the C-terminal region of human OXTR.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human hysteromyoma Tissue, 1:20-1:200

Type: Primary
Antigen: OXTR
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR