Anti-MAGOH Rabbit Polyclonal Antibody
Supplier: Proteintech
12347-1-AP-150UL
10089-906EA 709.42 CAD
10089-906
Anti-MAGOH Rabbit Polyclonal Antibody
Antibodies
MAGOH, belonging to the mago nashi family, is a component of a splicing-dependent multiprotein exon junction complex (EJC) deposited at splice junction on mRNAs. The EJC is a dynamic structure consisting of a few core proteins and several more peripheral nuclear and cytoplasmic associated factors that join the complex only transiently either during EJC assembly or during subsequent mRNA metabolism. Core components of the EJC functions to mark the position of the exon-exon junction in the mature mRNA and thereby influences downstream processes of gene expression including mRNA splicing,More Product Information