Anti-MMAB Rabbit Polyclonal Antibody

Supplier: Proteintech
11137-1-AP-150UL
10089-660EA 709.42 CAD
10089-660
Anti-MMAB Rabbit Polyclonal Antibody
Antibodies
The MMAB gene encodes cob(I)alamin adenosyltransferase, which catalyzes the final step in the synthesis of the coenzyme adenosylcobalamin (AdoCbl). AdoCbl is a vitamin B12-containing coenzyme for methylmalonyl-CoA mutase. The deduced 250-amino acid protein has a calculated molecular mass of 27.3 kD.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: MMAB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR