Anti-MAT2B Rabbit Polyclonal Antibody

Supplier: Proteintech
15952-1-AP-150UL
10089-656EA 709.42 CAD
10089-656
Anti-MAT2B Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human lung, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: MAT2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR