Anti-Kynurenine 3 monooxygenase Mouse Monoclonal Antibody [clone: 2D2F2]

Supplier: Proteintech
60029-1-IG-150UL
10089-348EA 709.42 CAD
10089-348
Anti-Kynurenine 3 monooxygenase Mouse Monoclonal Antibody [clone: 2D2F2]
Antibodies
KMO(Kynurenine 3-monooxygenase) is an NADPH-dependent flavin monooxygenase, catalysing the hydroxylation of the L-tryptophan metabolite L-kynurenine to form L-3-hydroxykynurenine. KMO is a membrane protein located on the outer membrane of mitochondria. Tissue distribution studies have revealed that, in rats, highest enzyme activity is found in kidney and liver, with brain having the least activity in comparison to peripheral organs.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Heart Tissue, 1:20-1:200; IF:C6 Cells, 1:10-1:100

Type: Primary
Antigen: Kynurenine 3 monooxygenase
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol Kynurenine 3 monooxygenase
Antigen name Kynurenine 3 monooxygenase
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 2D2F2
Reactivity Human,Rat,Mouse
Host Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Immunogen Recombinant Protein
Purification Caprylic acid/ammonium sulfate precipitation
Storage buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping temperature Store at -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR