Anti-GPRC5A Rabbit Polyclonal Antibody

Supplier: Proteintech
10309-1-AP-150UL
10087-804EA 709.42 CAD
10087-804
Anti-GPRC5A Rabbit Polyclonal Antibody
Antibodies
GPRC5A, also named as GPCR5A, RAI3 and RAIG1, belongs to the G-protein coupled receptor 3 family. It is an orphan G-protein coupled receptor (GPCR) that has been associated with malignancy and may play a role in the proliferation of breast cancer cells. GPRC5A is a potential molecular target for treatment of breast cancers. It is suppressed in some human lung carcinoma cells.

IHC: Human breast cancer Tissue, 1:20-1:200; Western Blot:Transfected HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: GPRC5A,RAI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol GPRC5A
Environmentally Preferable
Antigen name G protein-coupled Receptor Cass C Group 5 Member A
Antigen synonyms GPCR5A|RAIG1|RAI3|PEIG-1|TIG1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunogen Fusion Protein
Purification Antigen affinity Purification
Storage buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping temperature Store at -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR