Anti-GNRHR Rabbit Polyclonal Antibody
Supplier: Proteintech
19950-1-AP-150UL
10087-722EA 709.42 CAD
10087-722
Anti-GNRHR Rabbit Polyclonal Antibody
Antibodies
GNRHR, also named as GRHR, belongs to the G-protein coupled receptor 1 family. GNRHR is a receptor for gonadotropin releasing hormone (GnRH) that mediates the action of GnRH to stimulate the secretion of the gonadotropic hormones luteinizing hormone (LH) and follicle-stimulating hormone (FSH). It mediates its action by association with G-proteins that activate a phosphatidylinositol-calcium second messenger system. Isoform2 of GNRHR may act as an inhibitor of GnRH-R signaling. Defects in GNRHR are a cause of idiopathic hypogonadotropic hypogonadism (IHH). Defects in GNRHR are a cause ofMore Product Information