Anti-GOLGA2/GM130 Rabbit Polyclonal Antibody
Supplier: Proteintech
11308-1-AP-150UL
10087-264EA 709.42 CAD
10087-264
Anti-GOLGA2/GM130 Rabbit Polyclonal Antibody
Antibodies
GOLGA2, also known as GM130, is a 130 kDa cis-Golgi matrix protein which is one component of the detergent and salt resistant Golgi matrix. It is a peripheral membrane protein highly bound to Golgi membrane and localized mainly at the cytoplasmic face of cis-Golgi membrane. Together with its interacting partner proteins, including p115, giantin, GRASP65, and Rab GTPase, GOLGA2/GM130 is involved in the regulation of ER-to-Golgi transport and also in the maintenance of the Golgi structure. Emerging evidence suggest that the GOLGA2/GM130 has potential roles in the control of glycosylation, cellMore Product Information